5-(1,1-Difluoropropyl)-1,2,3-trifluorobenzene ≥95%

Supplier: Matrix Scientific

034830-1G
101838-454EA 380 USD
101838-454
5-(1,1-Difluoropropyl)-1,2,3-trifluorobenzene ≥95%
5-(1,1-Difluoropropyl)-1,2,3-trifluorobenzene

Hazard: Irritant

Formula: C₉H₇F₅
MDL Number: MFCD11845842
CAS Number: 1138445-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR