2-(1,1-Difluoropropyl)-1,3,4-trifluorobenzene ≥95%

Supplier: Matrix Scientific

034828-1G
101838-450EA 201.54 USD
101838-450
2-(1,1-Difluoropropyl)-1,3,4-trifluorobenzene ≥95%
2-(1,1-Difluoropropyl)-1,3,4-trifluorobenzene

Hazard: Irritant

Formula: C₉H₇F₅
MDL Number: MFCD11845840
CAS Number: 1138445-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR