1-(1,1-Difluoropropyl)-2,3-difluorobenzene ≥95%

Supplier: Matrix Scientific

034822-1G
101838-438EA 207.69 USD
101838-438
1-(1,1-Difluoropropyl)-2,3-difluorobenzene ≥95%
1-(1,1-Difluoropropyl)-2,3-difluorobenzene

Hazard: Irritant

Formula: C₉H₈F₄
MDL Number: MFCD11845836
CAS Number: 1138445-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR