4-(Difluorophenylmethyl)-2-fluoro-1-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

034816-1G
101838-426EA 209.23 USD
101838-426
4-(Difluorophenylmethyl)-2-fluoro-1-(trifluoromethyl)benzene ≥95%
4-(Difluorophenylmethyl)-2-fluoro-1-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₁₄H₈F₆
MDL Number: MFCD11845830
CAS Number: 1138445-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR