1-(Difluorophenylmethyl)-2-fluoro-3-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

034815-1G
101838-424EA 207.69 USD
101838-424
1-(Difluorophenylmethyl)-2-fluoro-3-(trifluoromethyl)benzene ≥95%
1-(Difluorophenylmethyl)-2-fluoro-3-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₁₄H₈F₆
MDL Number: MFCD11845829
CAS Number: 1138445-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR