2-(Difluorophenylmethyl)-1,4-difluorobenzene ≥95%

Supplier: Matrix Scientific

034813-1G
101838-420EA 153.85 USD
101838-420
2-(Difluorophenylmethyl)-1,4-difluorobenzene ≥95%
2-(Difluorophenylmethyl)-1,4-difluorobenzene

Hazard: Irritant

Formula: C₁₃H₈F₄
MDL Number: MFCD11845827
CAS Number: 1138445-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR