1-(Difluorophenylmethyl)-3,5-bis(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

034811-1G
101838-416EA 198.46 USD
101838-416
1-(Difluorophenylmethyl)-3,5-bis(trifluoromethyl)benzene ≥95%
1-(Difluorophenylmethyl)-3,5-bis(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₁₅H₈F₈
MDL Number: MFCD11845825
CAS Number: 1138445-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR