1-(1,1-Difluoroethyl)-3-(trifluoromethyl)benzene ≥95%
Supplier: Matrix Scientific
034809-1G
101838-412EA
144.62
USD
101838-412
1-(1,1-Difluoroethyl)-3-(trifluoromethyl)benzene ≥95%
1-(1,1-Difluoroethyl)-3-(trifluoromethyl)benzene
Hazard: Irritant
Formula:
C₉H₇F₅ |
MDL Number:
MFCD11845823 CAS Number: 1071212-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...