2-(1,1-Difluoroethyl)-1-fluoro-3-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

034803-1G
101838-400EA 207.69 USD
101838-400
2-(1,1-Difluoroethyl)-1-fluoro-3-(trifluoromethyl)benzene ≥95%
2-(1,1-Difluoroethyl)-1-fluoro-3-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₉H₆F₆
MDL Number: MFCD11845817
CAS Number: 1138445-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR