2-(1,1-Difluoroethyl)-1-fluoro-4-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

034801-1G
101838-396EA 203.08 USD
101838-396
2-(1,1-Difluoroethyl)-1-fluoro-4-(trifluoromethyl)benzene ≥95%
2-(1,1-Difluoroethyl)-1-fluoro-4-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₉H₆F₆
MDL Number: MFCD11226715
CAS Number: 1022980-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR