1-(1,1-Difluoroethyl)-2-fluoro-4-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

034798-1G
101838-390EA 209.23 USD
101838-390
1-(1,1-Difluoroethyl)-2-fluoro-4-(trifluoromethyl)benzene ≥95%
1-(1,1-Difluoroethyl)-2-fluoro-4-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₉H₆F₆
MDL Number: MFCD11226706
CAS Number: 1138445-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR