Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(1,1-Difluoroethyl)-3-fluorobenzene ≥95%

Supplier: Matrix Scientific


034796-1G
101838-386EA 130.77 USD InStock
101838-386
1-(1,1-Difluoroethyl)-3-fluorobenzene ≥95%
1-(1,1-Difluoroethyl)-3-fluorobenzene
Purity: 95%
CAS: 1138445-15-4
MDL Number: MFCD11845815
Molecular Formula: C8H7F3
Molecular Weight: 160,14
Hazard: Irritant


Formula: C₈H₇F₃
MW: 160,14 g/mol
MDL Number: MFCD11845815
CAS Number: 1138445-15-4
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty