4-(1,1-Difluoroethyl)-1,2-difluorobenzene ≥95%

Supplier: Matrix Scientific

034790-1G
101838-374EA 143.08 USD
101838-374
4-(1,1-Difluoroethyl)-1,2-difluorobenzene ≥95%
4-(1,1-Difluoroethyl)-1,2-difluorobenzene

Hazard: Irritant

Formula: C₈H₆F₄
MDL Number: MFCD11845811
CAS Number: 1138445-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR