2-(1,1-Difluoroethyl)-1,3-difluorobenzene ≥95%

Supplier: Matrix Scientific

034787-1G
101838-368EA 684.62 USD
101838-368
2-(1,1-Difluoroethyl)-1,3-difluorobenzene ≥95%
2-(1,1-Difluoroethyl)-1,3-difluorobenzene

Hazard: Irritant

Formula: C₈H₆F₄
MDL Number: MFCD11845808
CAS Number: 1138445-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR