2-(1,1-Difluoroethyl)-1,4-difluorobenzene ≥95%

Supplier: Matrix Scientific

034786-1G
101838-366EA 153.85 USD
101838-366
2-(1,1-Difluoroethyl)-1,4-difluorobenzene ≥95%
2-(1,1-Difluoroethyl)-1,4-difluorobenzene

Hazard: Irritant

Formula: C₈H₆F₄
MDL Number: MFCD11845807
CAS Number: 1138445-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR