1-(1,1-Difluoroethyl)-2,3-difluoro-4-methylbenzene ≥95%

Supplier: Matrix Scientific

034785-1G
101838-364EA 476.92 USD
101838-364
1-(1,1-Difluoroethyl)-2,3-difluoro-4-methylbenzene ≥95%
1-(1,1-Difluoroethyl)-2,3-difluoro-4-methylbenzene

Hazard: Irritant

Formula: C₉H₈F₄
MDL Number: MFCD11845806
CAS Number: 1138445-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR