1,4-Dichloro-2-(difluorophenylmethyl)benzene ≥95%

Supplier: Matrix Scientific

034783-1G
101838-360EA 176.92 USD
101838-360
1,4-Dichloro-2-(difluorophenylmethyl)benzene ≥95%
1,4-Dichloro-2-(difluorophenylmethyl)benzene

Hazard: Irritant

Formula: C₁₃H₈Cl₂F₂
MDL Number: MFCD11845805
CAS Number: 1138445-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR