2-Chloro-3-(1,1-difluoroethyl)-4-fluoro-1-methylbenzene ≥95%

Supplier: Matrix Scientific

034775-1G
101838-344EA 223.08 USD
101838-344
2-Chloro-3-(1,1-difluoroethyl)-4-fluoro-1-methylbenzene ≥95%
2-Chloro-3-(1,1-difluoroethyl)-4-fluoro-1-methylbenzene

Hazard: Irritant

Formula: C₉H₈ClF₃
MDL Number: MFCD11845799
CAS Number: 1138444-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR