1-Bromo-2-(difluorophenylmethyl)benzene ≥95%

Supplier: Matrix Scientific

034771-1G
101838-336EA 170.77 USD
101838-336
1-Bromo-2-(difluorophenylmethyl)benzene ≥95%
1-Bromo-2-(difluorophenylmethyl)benzene

Hazard: Irritant

Formula: C₁₃H₉BrF₂
MDL Number: MFCD11845797
CAS Number: 263249-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR