2-Bromo-1-(1,1-difluoroethyl)-4-fluorobenzene ≥95%

Supplier: Matrix Scientific

034770-1G
101838-334EA 90.77 USD
101838-334
2-Bromo-1-(1,1-difluoroethyl)-4-fluorobenzene ≥95%
2-Bromo-1-(1,1-difluoroethyl)-4-fluorobenzene

Hazard: Irritant

Formula: C₈H₆BrF₃
MDL Number: MFCD11845796
CAS Number: 1138444-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR