2,6-Dichloro-5-fluoro-3-(hydroxymethyl)pyridine ≥98%

Supplier: Matrix Scientific

034756-1G
101838-302EA 89.23 USD
101838-302
2,6-Dichloro-5-fluoro-3-(hydroxymethyl)pyridine ≥98%
2,6-Dichloro-5-fluoro-3-(hydroxymethyl)pyridine

Hazard: Irritant

Formula: C₆H₄Cl₂FNO
MDL Number: MFCD04972782
CAS Number: 820224-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR