5-Chloro-2,4-difluoropyrimidine ≥98%

Supplier: Matrix Scientific

034750-1G 034750-5G
101838-286EA 136.92 USD
101838-286 101838-288
5-Chloro-2,4-difluoropyrimidine ≥98%
5-Chloro-2,4-difluoropyrimidine

Hazard: Irritant

Formula: C₄HClF₂N₂
MDL Number: MFCD04972716
CAS Number: 25151-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR