6-Methyl-2,3-bis-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

034737-1G
101838-256EA 283.08 USD
101838-256
6-Methyl-2,3-bis-(trifluoromethyl)pyridine ≥95%
6-Methyl-2,3-bis-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₈H₅F₆N
MDL Number: MFCD11226677
CAS Number: 1099597-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR