2-(2,4-Dichlorobenzyl)thiourea hydrochloride ≥97%

Supplier: Matrix Scientific

034734-5G
101838-250EA 95.38 USD
101838-250
2-(2,4-Dichlorobenzyl)thiourea hydrochloride ≥97%
2-(2,4-Dichlorobenzyl)thiourea hydrochloride

Hazard: Irritant

Formula: C₈H₉Cl₃N₂S
MDL Number: MFCD11506648
CAS Number: 1185301-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR