1-Methyl-3-(3,3,3-trifluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

034726-1G
101838-234EA 347.69 USD
101838-234
1-Methyl-3-(3,3,3-trifluoropropyl)benzene ≥95%
1-Methyl-3-(3,3,3-trifluoropropyl)benzene

Hazard: Irritant

Formula: C₁₀H₁₁F₃
MDL Number: MFCD11226685
CAS Number: 1099597-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR