1-Methyl-2-(3,3,3-trifluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

034724-1G
101838-230EA 300 USD
101838-230
1-Methyl-2-(3,3,3-trifluoropropyl)benzene ≥95%
1-Methyl-2-(3,3,3-trifluoropropyl)benzene

Hazard: Irritant

Formula: C₁₀H₁₁F₃
MDL Number: MFCD11226684
CAS Number: 1099597-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR