1-Chloro-2-(3,3,3-trifluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

034720-1G
101838-220EA 255.38 USD
101838-220
1-Chloro-2-(3,3,3-trifluoropropyl)benzene ≥95%
1-Chloro-2-(3,3,3-trifluoropropyl)benzene

Hazard: Irritant

Formula: C₉H₈ClF₃
MDL Number: MFCD11226680
CAS Number: 96256-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR