2,3-Difluoro-4-methylbenzotrifluoride ≥95%

Supplier: Matrix Scientific

034716-1G
101838-212EA 256.92 USD
101838-212
2,3-Difluoro-4-methylbenzotrifluoride ≥95%
2,3-Difluoro-4-methylbenzotrifluoride

Hazard: Irritant

Formula: C₈H₅F₅
MDL Number: MFCD03701050
CAS Number: 1099597-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR