1-Chloro-2-(2,2,2-trifluoroethyl)-4-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

034715-1G
101838-210EA 283.08 USD
101838-210
1-Chloro-2-(2,2,2-trifluoroethyl)-4-(trifluoromethyl)benzene ≥95%
1-Chloro-2-(2,2,2-trifluoroethyl)-4-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₉H₅ClF₆
MDL Number: MFCD11226654
CAS Number: 1099597-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR