1,2,3,4,5-Pentafluoro-6-(3,3,3-trifluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

034706-1G
101838-192EA 224.62 USD
101838-192
1,2,3,4,5-Pentafluoro-6-(3,3,3-trifluoropropyl)benzene ≥95%
1,2,3,4,5-Pentafluoro-6-(3,3,3-trifluoropropyl)benzene

Hazard: Irritant

Formula: C₉H₄F₈
MDL Number: MFCD11226671
CAS Number: 1099597-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR