4-Fluoro-1-(2,2,2-trifluoroethyl)-2-(trifluoromethyl)benzene

Supplier: Matrix Scientific

034702-1G
101838-184EA 303.08 USD
101838-184
4-Fluoro-1-(2,2,2-trifluoroethyl)-2-(trifluoromethyl)benzene
4-Fluoro-1-(2,2,2-trifluoroethyl)-2-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₉H₅F₇
MDL Number: MFCD11226670
CAS Number: 1099597-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR