4-Fluoro-2-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

034700-1G
101838-180EA 364.62 USD
101838-180
4-Fluoro-2-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene ≥95%
4-Fluoro-2-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₉H₅F₇
MDL Number: MFCD11226669
CAS Number: 1099597-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR