5-(2,2,2-Trifluoroethyl)octahydrobenzofuran ≥95%

Supplier: Matrix Scientific

034699-1G
101838-178EA 218.46 USD
101838-178
5-(2,2,2-Trifluoroethyl)octahydrobenzofuran ≥95%
5-(2,2,2-Trifluoroethyl)octahydrobenzofuran

Hazard: Irritant

Formula: C₁₀H₁₅F₃O
MDL Number: MFCD11226646
CAS Number: 1212378-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR