4-Chloro-2-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene ≥95%
Supplier: Matrix Scientific
034698-1G
101838-176EA
267.69
USD
101838-176
4-Chloro-2-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene ≥95%
4-Chloro-2-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene
Hazard: Irritant
Formula:
C₉H₅ClF₆ |
MDL Number:
MFCD11226668 CAS Number: 1099597-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...