1,2-Dimethyl-4-(3,3,3-trifluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

034689-1G
101838-158EA 396.92 USD
101838-158
1,2-Dimethyl-4-(3,3,3-trifluoropropyl)benzene ≥95%
1,2-Dimethyl-4-(3,3,3-trifluoropropyl)benzene

Hazard: Irritant

Formula: C₁₁H₁₃F₃
MDL Number: MFCD11226688
CAS Number: 1099597-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR