2,4-Dimethyl-1-(3,3,3-trifluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

034686-1G
101838-152EA 300 USD
101838-152
2,4-Dimethyl-1-(3,3,3-trifluoropropyl)benzene ≥95%
2,4-Dimethyl-1-(3,3,3-trifluoropropyl)benzene

Hazard: Irritant

Formula: C₁₁H₁₃F₃
MDL Number: MFCD11226687
CAS Number: 1099597-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR