2-Fluoro-1-(2,2,2-trifluoroethyl)-3-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

034685-1G
101838-150EA 364.62 USD
101838-150
2-Fluoro-1-(2,2,2-trifluoroethyl)-3-(trifluoromethyl)benzene ≥95%
2-Fluoro-1-(2,2,2-trifluoroethyl)-3-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₉H₅F₇
MDL Number: MFCD11226660
CAS Number: 1099597-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR