4-Bromo-1-ethyl-1H-pyrazole

Supplier: Matrix Scientific

Synonyms: 4-Bromo-1-ethylpyrazole

034681-500MG
101838-142EA 29.9 USD
101838-142
4-Bromo-1-ethyl-1H-pyrazole
4-Bromo-1-ethyl-1H-pyrazole

Hazard: Irritant

Formula: C₅H₇BrN₂
MW: 175.0265 g/mol
MDL Number: MFCD08235252
CAS Number: 71229-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR