4-Bromo-1-ethyl-5-methyl-1H-pyrazole

Supplier: Matrix Scientific

034679-500MG
101838-136EA 307.69 USD
101838-136
4-Bromo-1-ethyl-5-methyl-1H-pyrazole
4-Bromo-1-ethyl-5-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₆H₉BrN₂
MDL Number: MFCD11500994
CAS Number: 1171667-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR