C-(4-Methyl-[1,2,3]thiadiazol-5-yl)methylamine

Supplier: Matrix Scientific

034672-500MG
101838-122EA 307.69 USD
101838-122
C-(4-Methyl-[1,2,3]thiadiazol-5-yl)methylamine
C-(4-Methyl-[1,2,3]thiadiazol-5-yl)methylamine

Hazard: Irritant

Formula: C₄H₇N₃S
MDL Number: MFCD10001570
CAS Number: 790641-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR