4-Isopropyl-1,2,3-thiadiazole-5-carboxylic acid

Supplier: Matrix Scientific

034670-500MG 034670-1G
101838-118EA 469.23 USD
101838-118 101838-116
4-Isopropyl-1,2,3-thiadiazole-5-carboxylic acid
4-Isopropyl-1,2,3-thiadiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₆H₈N₂O₂S
MDL Number: MFCD01935987
CAS Number: 183302-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR