1-Ethyl-4-(3,3,3-trifluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

032561-1G
101834-000EA 267.69 USD
101834-000
1-Ethyl-4-(3,3,3-trifluoropropyl)benzene ≥95%
1-Ethyl-4-(3,3,3-trifluoropropyl)benzene

Hazard: Irritant

Formula: C₁₁H₁₃F₃
MDL Number: MFCD11226586
CAS Number: 1099597-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR