1-Isopropyl-4-(3,3,3-trifluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

032558-1G
101833-994EA 267.69 USD
101833-994
1-Isopropyl-4-(3,3,3-trifluoropropyl)benzene ≥95%
1-Isopropyl-4-(3,3,3-trifluoropropyl)benzene

Hazard: Irritant

Formula: C₁₂H₁₅F₃
MDL Number: MFCD11226585
CAS Number: 1099597-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR