1-Fluoro-2,3-bis(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

032557-1G
101833-992EA 250.77 USD
101833-992
1-Fluoro-2,3-bis(trifluoromethyl)benzene ≥95%
1-Fluoro-2,3-bis(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₃F₇
MW: 232.1 g/mol
MDL Number: MFCD11226555
CAS Number: 1099597-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR