1,4-Difluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

032548-1G
101833-970EA 224.62 USD
101833-970
1,4-Difluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%
1,4-Difluoro-2-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₅F₅
MDL Number: MFCD11226583
CAS Number: 1099597-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR