2-Chloro-4-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

032547-1G
101833-968EA 240 USD
101833-968
2-Chloro-4-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene ≥95%
2-Chloro-4-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₉H₅ClF₆
MDL Number: MFCD11226553
CAS Number: 1099597-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR