4-Fluoro-3-methylbenzotrifluoride ≥95%

Supplier: Matrix Scientific

032544-1G
101833-962EA 221.54 USD
101833-962
4-Fluoro-3-methylbenzotrifluoride ≥95%
4-Fluoro-3-methylbenzotrifluoride

Hazard: Irritant

Formula: C₈H₆F₄
MDL Number: MFCD07778352
CAS Number: 74483-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR