4-(1-Hydroxyimino-2-methylpropylcarbamoyl)piperidine-1-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

032530-1G
101833-930EA 138.46 USD
101833-930
4-(1-Hydroxyimino-2-methylpropylcarbamoyl)piperidine-1-carboxylic acid tert-butyl ester ≥95%
4-(1-Hydroxyimino-2-methylpropylcarbamoyl)piperidine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₅H₂₇N₃O₄
MW: 313.4 g/mol
MDL Number: MFCD11226633
CAS Number: 913264-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR