2-Bromo-N-(5-chloro-2-hydroxyphenyl)-2-methylpropionamide ≥95%

Supplier: Matrix Scientific

032522-1G
101833-916EA 127.69 USD
101833-916
2-Bromo-N-(5-chloro-2-hydroxyphenyl)-2-methylpropionamide ≥95%
2-Bromo-N-(5-chloro-2-hydroxyphenyl)-2-methylpropionamide

Hazard: Irritant

Formula: C₁₀H₁₁BrClNO₂
Melting Pt: 172 °C
MDL Number: MFCD11226533
CAS Number: 327026-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR