4-Chloro-1-fluoro-2-(2,2,2-trifluoroethyl)benzene ≥97%

Supplier: Matrix Scientific

032516-1G
101833-902EA 476.92 USD
101833-902
4-Chloro-1-fluoro-2-(2,2,2-trifluoroethyl)benzene ≥97%
4-Chloro-1-fluoro-2-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₅ClF₄
MDL Number: MFCD11226523
CAS Number: 1099598-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR